Xylenes, Semiconductor Grade
Xylenes, Semiconductor Grade
Xylenes, Semiconductor Grade
Xylenes, Semiconductor Grade
Thermo Scientific Chemicals
Xylenes, Semiconductor Grade
CAS: 1330-20-7 | (CH3)2C6H4
Have Questions?
Catalog NumberQuantity
FSK019402.K21 L
Catalog number FSK019402.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialXylenes
Melting Point<°C to 25°C
CAS1330-20-7
ColorColorless
Health Hazard 1H226-H304-H312+H332-H315-H319-H335-H373
View more
Xylene is used as a solvent in pharmaceutical, printing, rubber, and leather industries. It is an important component of ink, rubber and adhesives as well as in thinning paints and varnishes. It is used in the laboratory to make baths with dry ice to cool reaction vessels, and as a solvent to remove synthetic immersion oil from the microscope objective in light microscopy. It finds application as a short-term hydrophobic clearing agent of tissue samples by histologists. It is a precursor for the preparation of xylene dichlorides, xylyl bromide, terephthalic acid and dimethyl terephthalate. It acts as a cleaning agent for steel, silicon wafers and integrated circuits.