Potassium acetate, ACS, 99% min
Potassium acetate, ACS, 99% min
Potassium acetate, ACS, 99% min
Potassium acetate, ACS, 99% min
Potassium acetate, ACS, 99% min
Thermo Scientific Chemicals
Potassium acetate, ACS, 99% min
CAS: 127-08-2 | C2H3KO2 | 98.142 g/mol
Have Questions?
Catalog NumberQuantity
FSK013449.22100 g
Catalog number FSK013449.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium acetate
Melting Point292°C
Name NoteFor ACS analysis
CAS127-08-2
Health Hazard 1H500
View more
Potassium acetate is used as deicer to remove snow and ice from the surface and is considered as a replacement for calcium chloride or magnesium chloride. It is used as a food additive; as a buffer in biological research; as extinguishing agent used in class K fire extinguishers and as a catalyst to produce polyurethane polymers. It is utilized for the precipitation of dodecyl sulfate bound proteins thereby removal of proteins from DNA. It plays an important role for preserving tissues, fixation and mummification. .