Potassium nitrate, ACS, 99.0% min
Potassium nitrate, ACS, 99.0% min
Potassium nitrate, ACS, 99.0% min
Potassium nitrate, ACS, 99.0% min
Thermo Scientific Chemicals
Potassium nitrate, ACS, 99.0% min
For Pyrotechnics, Glass Manufacture, Tempering Steel | CAS: 7757-79-1 | KNO3 | 101.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number FSK013443.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Boiling Point400°C (decomposition)
Chemical Name or MaterialPotassium nitrate
Melting Point334°C
CAS7757-79-1
ColorWhite
View more
Pyrotechnics, glass manufacture, tempering steel, reagentPotassium nitrate has a vast variety of applications. Appropriate preparations of potassium nitrate find use in fertilizers, food preservatives, tooth paste, rocket propellants and fireworks. In addition, it is also used as an electrolyte in a salt bridge, and as thermal storage medium in power generation systems. It forms phase change material with silica and alumina nanoparticles. In combination with boron trifluoride it forms an efficient reagent for nitration of aromatic compounds. Adsorption on alumina provides an environmentally benign aromatic nitrating agent.