Rubidium bromide, 99.8% (metals basis)
Rubidium bromide, 99.8% (metals basis)
Rubidium bromide, 99.8% (metals basis)
Rubidium bromide, 99.8% (metals basis)
Thermo Scientific Chemicals
Rubidium bromide, 99.8% (metals basis)
CAS: 7789-39-1 | BrRb | 165.37 g/mol
Have Questions?
Catalog NumberQuantity
FSK012891.065 g
Catalog number FSK012891.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Boiling Point1340°C
Chemical Name or MaterialRubidium bromide
Melting Point693°C
CAS7789-39-1
Recommended StorageAmbient temperatures
View more
Rubidium bromide is used in infrared optics.