Zinc oxide, Puratronic™, 99.9995% (metals basis)
Zinc oxide, Puratronic™, 99.9995% (metals basis)
Zinc oxide, Puratronic™, 99.9995% (metals basis)
Zinc oxide, Puratronic™, 99.9995% (metals basis)
Thermo Scientific Chemicals
Zinc oxide, Puratronic™, 99.9995% (metals basis)
CAS: 1314-13-2 | OZn | 81.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number FSK012855.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialZinc oxide
Melting Point1975°C
CAS1314-13-2
ColorWhite
Recommended StorageAmbient temperatures
View more
Zinc oxide is used as a sun screener. It is serves as an additive in products such as cigarette filters, plastics, anticorrosive coatings, ceramics, glass, cement, lubricants, ointments, adhesives, sealants, pigments, batteries, ferrites, fire retardants, and first-aid tapes. Zinc oxide plays an important role in textiles as UV-absorber. It can protect rubber from fungi and UV light. As a wide-bandgap semiconductor with favorable properties, it has the potential for use in transparent electrodes, in nanorod sensors, in liquid crystal displays, as thin-film transistors and light emitting diodes, and in heat-protecting windows.