Sodium chloride, ACS, 99.0% min
Sodium chloride, ACS, 99.0% min
Sodium chloride, ACS, 99.0% min
Sodium chloride, ACS, 99.0% min
Thermo Scientific Chemicals
Sodium chloride, ACS, 99.0% min
CAS: 7647-14-5 | ClNa | 58.44 g/mol
Have Questions?
Catalog NumberQuantity
FSK012314.36500 g
Catalog number FSK012314.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Boiling Point1413°C
Chemical Name or MaterialSodium chloride
Melting Point801°C
Name NoteFor analysis ACS
CAS7647-14-5
View more
Sodium chloride is widely used as a food additive, and a preservative. It is employed in the Chloralkali process for producing chlorine and sodium hydroxide, in the Solvay process for producing sodium carbonate and calcium chloride, in the Mannheim process and in the Hargreaves process for producing sodium sulfate and hydrochloric acid. It is used in the manufacture of sodium chlorate, water softening and de-icing of roadways. Further it is used in several industries like pharmaceutical, textile, dyeing, rubber food and agriculture. It is used in the processing of steel, copper, vanadium, and other metals.