Silver foil, 0.1mm (0.004in) thick, hard, Premion™, 99.998% (metals basis)
Silver foil, 0.1mm (0.004in) thick, hard, Premion™, 99.998% (metals basis)
Silver foil, 0.1mm (0.004in) thick, hard, Premion™, 99.998% (metals basis)
Silver foil, 0.1mm (0.004in) thick, hard, Premion™, 99.998% (metals basis)
Thermo Scientific Chemicals
Silver foil, 0.1mm (0.004in) thick, hard, Premion™, 99.998% (metals basis)
Form: Approx. 2.6g/50 x 50mm; Temper: as rolled | CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Catalog NumberQuantity
FSK012126.HP100 x 500 mm
Catalog number FSK012126.HP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 x 500 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialSilver
ColorSilver
Name NoteHard
CAS7440-22-4
AppearanceSilver-colored foil