Sodium phosphate dodecahydrate, ACS, 98.0-102.0%
Sodium phosphate dodecahydrate, ACS, 98.0-102.0%
Sodium phosphate dodecahydrate, ACS, 98.0-102.0%
Sodium phosphate dodecahydrate, ACS, 98.0-102.0%
Thermo Scientific Chemicals
Sodium phosphate dodecahydrate, ACS, 98.0-102.0%
CAS: 10101-89-0 | H24Na3O16P | 380.119 g/mol
Have Questions?
Catalog NumberQuantity
FSK011590.36500 g
Catalog number FSK011590.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium phosphate dodecahydrate
Melting Point∼75°C (decomposition)
CAS10101-89-0
ColorWhite
Health Hazard 1H315-H319-H335
View more
Sodium phosphate dodecahydrate is used in photographic developing, clarifying sugar, removing boiler scale, softening water and manufacturing of paper. They are added to many foods as an emulsifier to prevent oil separation. It can be utilized as a leavening agent. Some instances of these foods include the batter coating on breaded fish or chicken, and commercially baked cakes. Adding this item to food increases the shelf life of the food, maintaining the texture and visual aspect of the food.