Tin(IV) chloride, anhydrous, 98% (metals basis)
Tin(IV) chloride, anhydrous, 98% (metals basis)
Tin(IV) chloride, anhydrous, 98% (metals basis)
Tin(IV) chloride, anhydrous, 98% (metals basis)
Thermo Scientific Chemicals
Tin(IV) chloride, anhydrous, 98% (metals basis)
CAS: 7646-78-8 | Cl4Sn | 260.51 g/mol
Have Questions?
Catalog NumberQuantity
FSK011571.36500 g
Catalog number FSK011571.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Boiling Point114°C
Chemical Name or MaterialTin(IV) chloride
Melting Point-33°C
Name NoteAnhydrous
CAS7646-78-8
View more
Tin(IV) chloride is a precursor to prepare organotin compounds such as tetralkyltin and dialkyldichlorotin(IV), which find applications as catalysts and polymer stabilizers. As a Lewis acid catalyst, it is used in Fridel-Crafts reactions for alkylation and cyclization. It is involved in the selective nitration of aromatic compounds in the presence of fuming nitric acid. Furthermore, it is used to prepare tin(IV) oxide coating by sol-gel process.