Manganese(II) chloride tetrahydrate, 99% (metals basis)
Manganese(II) chloride tetrahydrate, 99% (metals basis)
Manganese(II) chloride tetrahydrate, 99% (metals basis)
Manganese(II) chloride tetrahydrate, 99% (metals basis)
Thermo Scientific Chemicals
Manganese(II) chloride tetrahydrate, 99% (metals basis)
CAS: 13446-34-9 | Cl2H8MnO4 | 197.90 g/mol
Have Questions?
Catalog NumberQuantity
FSK011563.22100 g
Catalog number FSK011563.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialManganese(II) chloride tetrahydrate
Melting Point58°C
CAS13446-34-9
ColorPink
Health Hazard 1H302-H318-H373
View more
Manganese(II) chloride tetrahydrate is used in the tailing of DNA - MnCl2 buffer is recommended for dC and dG tailing and in reverse transcription of RNA with thermostable DNA polymerase.