Sodium acetate trihydrate, ACS, 99.0%-101%
Sodium acetate trihydrate, ACS, 99.0%-101%
Sodium acetate trihydrate, ACS, 99.0%-101%
Sodium acetate trihydrate, ACS, 99.0%-101%
Thermo Scientific Chemicals
Sodium acetate trihydrate, ACS, 99.0%-101%
CAS: 6131-90-4 | C2H9NaO5 | 136.079 g/mol
Have Questions?
Catalog NumberQuantity
FSK011553.36500 g
Catalog number FSK011553.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium Acetate Trihydrate
Melting Point62°C to 64°C
Name NoteFor ACS analysis
CAS6131-90-4
Health Hazard 1H500
View more
In photography, analytical laboratory as buffer and in pharmaceutical.Sodium acetate trihydrate is used in various applications. It is used as a buffer of a pH range in between to 4.0 to 6.0. It is also used in the purification and precipitation of nucleic acids and in protein crystallization. It is used as a mordant dyeing, as a pickling agent in chrome tanning, as a neutralizer in waste streams containing sulfuric acid, in the textile industry and as a photoresist while using aniline dyes. It plays an important role to retard vulcanization of chloroprene in synthetic rubber production. It is a precursor to prepare ester from alkyl halide.