Niobium(V) oxide, Puratronic™, 99.9985% (metals basis)
Niobium(V) oxide, Puratronic™, 99.9985% (metals basis)
Niobium(V) oxide, Puratronic™, 99.9985% (metals basis)
Niobium(V) oxide, Puratronic™, 99.9985% (metals basis)
Thermo Scientific Chemicals
Niobium(V) oxide, Puratronic™, 99.9985% (metals basis)
CAS: 1313-96-8 | Nb2O5 | 265.81 g/mol
Have Questions?
Catalog NumberQuantity
FSK010822.A11 kg
Catalog number FSK010822.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialNiobium(V) oxide
Melting Point1520°C
CAS1313-96-8
ColorWhite
Health Hazard 1H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
View more
Niobium pentoxide is the precursor to a majority of niobium compounds, powder metallurgy products, and alloys. It is also used in glass, optic and ceramic applications. The nanoparticles of niobium pentoxide are useful in the preparation of electrochemical capacitors and cathodes. In addition, appropriate preparations of this compound can be used as a dietary supplement in food industry. It is a precursor to lithium niobate which finds extensive use in piezoelectric sensors, anti-aliasing filters, optical waveguides, mobile phones, surface acoustic devices, optical modulators, including linear and non-linear optical applications.