Aluminum shot, approx. 4-8mm (0.2-0.3in), Puratronic™, 99.999% (metals basis)
Aluminum shot, approx. 4-8mm (0.2-0.3in), Puratronic™, 99.999% (metals basis)
Aluminum shot, approx. 4-8mm (0.2-0.3in), Puratronic™, 99.999% (metals basis)
Aluminum shot, approx. 4-8mm (0.2-0.3in), Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals
Aluminum shot, approx. 4-8mm (0.2-0.3in), Puratronic™, 99.999% (metals basis)
approx. 4-8mm (0.2-0.3 in.) | CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Catalog NumberQuantity
FSK010573.A11 kg
Catalog number FSK010573.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialAluminum shot
Name Noteshot
CAS7429-90-5
Recommended StorageAmbient temperatures
RTECS NumberBD0330000