Ruthenium(III) 2,4-pentanedionate, Ru 24% min
Ruthenium(III) 2,4-pentanedionate, Ru 24% min
Ruthenium(III) 2,4-pentanedionate, Ru 24% min
Ruthenium(III) 2,4-pentanedionate, Ru 24% min
Thermo Scientific Chemicals
Ruthenium(III) 2,4-pentanedionate, Ru 24% min
For Hydrogenation | CAS: 14284-93-6 | C15H21O6Ru | 398.40 g/mol
Have Questions?
Catalog NumberQuantity
FSK010568.031 g
Catalog number FSK010568.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialRuthenium(III) 2, 4-pentanedionate
Melting Point260°C (decomposition)
Name NoteRu 24% min.
CAS14284-93-6
ColorWhite
View more
Precursor for the preparation of other compounds of ruthenium. Ruthenium(III) 2,4-pentanedionate is employed as a recyclable catalyst for certain organic transformations like the acetylation of phenols, alcohols, and amines under neat conditions. It is also used as a homogeneous catalyst, for example, in the hydrolysis of sodium borohydride and regiospecific tritiation of aromatic carboxylic acids. It is also used as a catalyst for enantioselective hydrogenation of acids such as aryl acrylic acid and aryl propenic acid.