▸
Sonicators
▸
Ultrasonic Homogenizers
▸
505 Sonicator with Probe
Actual product may vary
Thermo Scientific
505 Sonicator with Probe
Use this high powered sonicator for various applications and sample volumes. Fisherbrand™ Sonicators with Probe are ideal for cell disruption, nanoparticle dispersion, homogenisation and sample preparation.
Have Questions?
Catalog Number
Hertz
FQSFB505EUK-220
20 kHz
Catalog number
FQSFB505EUK-220
Price (SGD)
6,661.00
Special offer
Ends:
06-Sep-2026
9,516.00
Save 2,855.00 (30%)
Each
Add to cart
Hertz:
20 kHz
Price (SGD)
6,661.00
Special offer
Ends:
06-Sep-2026
9,516.00
Save 2,855.00 (30%)
Each
Add to cart
500W
Programmable from 1 second to 10 hours
Overload protection
Amplitude control from 20 to 100%
Processing volume 0.2 to 500mL
Specifications
Hertz
20 kHz
Wattage
500 W
Weight (Metric)
7 kg