Stand and Clamp for Model 50 and 120 Sonic Dismembrator
Stand and Clamp for Model 50 and 120 Sonic Dismembrator
Actual product may vary
Stand and Clamp for Model 50 and 120 Sonic Dismembrator
Stand and Clamp for Model 50 and 120 Sonic Dismembrator
Stand and Clamp for Model 50 and 120 Sonic Dismembrator
Stand and Converter clamp accessory for Model 50/120 Sonic Dismembrator. This accessory will properly support the converter and probe assembly during sonication.
Have Questions?
Catalog number FQSFB460
Price (SGD)
264.00
EA
Add to cart
Price (SGD)
264.00
EA
Add to cart
This Fisher Scientific™ Stand is designed for use with Model 50 and Model 120 Sonic Dismembrators
  • Laboratory Support Stand with Converter clamp
  • For use with Model 50/120 Sonic Dismembrator
Specifications
Product TypeStand
Height (English)24 in.
Height (Metric)61 cm
For Use With (Equipment)Model 50 and 120 sonic dismembrator
Includes0.5 in. dia. support rod and converter clamp
Unit SizeEA