Stand and Converter clamp for Model 505/705 Sonic Dismembrator. This accessory will properly support the converter and probe assembly during sonication.
Have Questions?
Catalog Number
For Use With (Equipment)
FQSFB459
Model 505 and 705 Sonic Dismembrator
Catalog number FQSFB459
Price (SGD)
182.00
Special offer
Ends: 06-Sep-2026
260.00
Save 78.00 (30%)
Each
For Use With (Equipment):
Model 505 and 705 Sonic Dismembrator
Price (SGD)
182.00
Special offer
Ends: 06-Sep-2026
260.00
Save 78.00 (30%)
Each
This Fisher Scientific™ Stand is designed for use with Model 505 and Model 705 Sonic Dismembrators
Laboratory Support Stand with Converter Clamp
For use with Model 505/705 Sonic Dismembrator
Specifications
Product TypeStand
Height (English)24 in.
Height (Metric)61 cm
For Use With (Equipment)Model 505 and 705 Sonic Dismembrator
Includes0.5 in. dia. support rod and converter clamp