Sound Enclosure for Models 505/705 Sonic Dismembrators
Actual product may vary
Sound Enclosure for Models 505/705 Sonic Dismembrators
Sound Enclosure for Model 505/705 Sonic Dismembrator. Sonic Dismembrators are extremely loud devices that can cause discomfort to the user and anyone nearby. The Sound Enclosure reduces noise by approximately 20 dBa and is made to work with all accessories. In addition to reducing noise, the Sound Enclosure has an internal support rod and converter mounting system. Any Fisherbrand probe or horn will be held safely and securely inside the unit.
Have Questions?
Catalog NumberLength (Metric)
FQSFB432B233 mm
Catalog number FQSFB432B2
Price (SGD)
1,730.00
Special offer
Ends: 06-Sep-2026
2,384.00
Save 654.00 (27%)
Each
Length (Metric):
33 mm
Price (SGD)
1,730.00
Special offer
Ends: 06-Sep-2026
2,384.00
Save 654.00 (27%)
Each
This Fisher Scientific™ Sound Enclosure is designed for use with Models 505/705 Sonic Dismembrators. Significantly reduces noise of Sonic Dismembrators.
  • Sound Reducing Enclosure including stand and clamp
  • For use with Model 505/705 Sonic Dismembrator
  • Two ports are located on either side of the enclosure for coolant tubing or a temperature monitoring probe
  • Interior walls are lined with water-resistant acoustical foam
  • Door has a window so experiments can be visually monitored
  • Dimensions: 13D x 13.5W x 30.5H in. (33 x 34.3 x 77.5cm)
Specifications
Product TypeSound Enclosure
Height (English)30.5 in.
Height (Metric)77.5 cm
Length (English)13 in.
Length (Metric)33 mm
Width (English)13.5 in.
Width (Metric)34.3 cm
For Use With (Application)Ideal for sound reduction
For Use With (Equipment)Model 505 and 705 sonic dismembrator
IncludesInternal support rod and converter mounting system
Unit SizeEach