Replaceable Tip Probe for use with 500 and 700 watt Sonic Dismembrators
Replaceable Tip Probe for use with 500 and 700 watt Sonic Dismembrators
Actual product may vary
Replaceable Tip Probe for use with 500 and 700 watt Sonic Dismembrators
Microtips are thin, high intensity probes which are designed for processing small sample volumes. Microtips screw into the threaded end of the standard 1/2 in. probe (4220).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) TipVolume Range (Metric)Includes
FQSFB422013 mm20 to 250 mLStandard 1/2 in. Probe
FQSFB420819 mm50 to 500 mLProbe
Catalog number FQSFB4220
Price (SGD)
1,393.00
EA
Add to cart
Diameter (Metric) Tip:
13 mm
Volume Range (Metric):
20 to 250 mL
Includes:
Standard 1/2 in. Probe
Price (SGD)
1,393.00
EA
Add to cart
These Fisher Scientific™ Probes are designed for use with 500 and 700 watt Sonic Dismembrators.
  • Available in diameters ranging from 1/16 in. to 1 in.
  • Capacity options of 0.2-5mL to 100-1000mL
  • Replaceable tip, solid and microtip probes
Specifications
Product TypeProbe
Amplitude120 μm maximum
Volume Range (Metric)20 to 250 mL
Diameter (English)0.5 in.
Diameter (English) Tip0.5 in.
Diameter (Metric)1.27 cm
Diameter (Metric) Tip13 mm
For Use With (Equipment)Model 505/705 Sonic dismembrator
IncludesStandard 1/2 in. Probe
Tip StyleReplaceable tip
Unit SizeEA