UltraSOLV™ Sponge
UltraSOLV™ Sponge
Actual product may vary
UltraSOLV™ Sponge
For use with UltraSOLV™ ScrubPADs on wet cleans
Have Questions?
Catalog NumberDimensions (L x W x H)
FOAHT47545.37 x 3.62 x 0.37 in.
Catalog number FOAHT4754
Price (SGD)
1,208.00
PK
Add to cart
Dimensions (L x W x H):
5.37 x 3.62 x 0.37 in.
Price (SGD)
1,208.00
PK
Add to cart
Fiber-free hydrophilic polyurethane foam construction combined with low levels of particle generation, ionics, and organics make these wipers ideal for for wet cleaning of process equipment in the semiconductor, disk drive, and flat-panel display industries.
  • Unique abrasion resistance for cleaning rough, coarse, and bead-blasted sufaces
  • Offers static dissipative and tribocharging properties
  • For static sensitive equipment
  • ISO Class 4 (Class 10) compatible
Specifications
Dimensions (L x W x H)5.37 x 3.62 x 0.37 in.
For Use With (Application)Performing wet cleans and PMs on vacuum based process equipment in the semiconductor, disk drive and flat panel display industries
IncludesStatic dissipative and tribocharging properties
Length (English)5.37 in.
Length (Metric)13.7 cm
MaterialPolyurethane Foam
Thickness (English)0.37 in.
Unit SizePK