UltraSORB™ Wipers
UltraSORB™ Wipers
Actual product may vary
UltraSORB™ Wipers
Open-cell, soft foam structure for class ISO 5 (Class 100) applications
Have Questions?
Catalog NumberDimensions (L x W)
FOAHT46999 x 9 in. (22.86 x 22.86 cm)
Catalog number FOAHT4699
Price (SGD)
58.00
PK
Add to cart
Dimensions (L x W):
9 x 9 in. (22.86 x 22.86 cm)
Price (SGD)
58.00
PK
Add to cart
Features unique pore structure allows for controlled solvent transfer to screen and metal masks to aid in removal of solder paste, conductive adhesives and thick film inks.
  • High density construction and sponge-like properties
  • Dry and low-moisture cleaning
  • Ideal for screen or stencil cleaning as well as medical device manufacturing operations
  • Static dissipative to minimize ESD risk
  • Pyrogen-free for wipe-down of medical devices such as catheters
  • Features very low ionic and organic signatures
  • 100% fiber free and abrasion resistant
Specifications
Product TypeUltraSORB™ Wipes
Length (English)9 in.
Length (Metric)22.86 cm
Width (English)9 in.
Width (Metric)22.86 cm
Dimensions (L x W)9 x 9 in. (22.86 x 22.86 cm)
For Use With (Application)For class ISO 5 (Class 100) applications, for screen or stencil cleaning as well as medical device manufacturing operations
For Use With (Equipment)For cleaning Metal masks used to print LTCC components, capacitors, EL, OLED, and plasma display panels
Thickness (Metric)0.27 cm
Unit SizePK