Barnstead™ 1/2 Size B-Pure™ Filters
Barnstead™ 1/2 Size B-Pure™ Filters
Actual product may vary
Barnstead™ 1/2 Size B-Pure™ Filters
Pre-treat your feed water with Thermo Scientific™ Barnstead™ 1/2 Size B-Pure™ Filters, available in varying pore sizes.
Have Questions?
Catalog NumberFor Use With (Equipment)TypePore Size
FL583X11/2 Size B-Pure Filter HoldersPrefilter5 μm
Catalog number FL583X1
Price (SGD)
155.00
EA
Add to cart
For Use With (Equipment):
1/2 Size B-Pure Filter Holders
Type:
Prefilter
Pore Size:
5 μm
Price (SGD)
155.00
EA
Add to cart
Pre-treat your feed water with Thermo Scientific™ Barnstead™ 1/2 Size B-Pure™ Filters. In addition to pre-treating feed water, the filters can act as a final filter to ensure consistently pure water. Compatible with B-Pure 1/2 size filter holders, these filters are available in varying pore sizes to meet all of your pre-filtration needs.
  • Large surface area for long life
  • Flow rates of up to 2L/min.
Specifications
Feedwater SourcePretreated
Flow RateUp to 2 L/min.
Pore Size5 μm
For Use With (Equipment)1/2 Size B-Pure Filter Holders
TypePrefilter
Length (English)10 in.
Length (Metric)25.4 cm
Diameter (English)2.5 in.
Diameter (Metric)6.5 cm
Unit SizeEA