Ultra Thin Polyester Laboratory Labels
Actual product may vary
Ultra Thin Polyester Laboratory Labels
Ultra Thin Polyester Laboratory Labels
Ultra Thin Polyester Laboratory Labels
Fisherbrand
Ultra Thin Polyester Laboratory Labels
Durable Freezerbondz™ Polyester Markers are designed for laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
Have Questions?
Catalog NumberProduct TypeSize
FKS17084377Label/Cartridge12.7 mm dia.
Catalog number FKS17084377
Price (SGD)
-
Product Type:
Label/Cartridge
Size:
12.7 mm dia.
B-492 labels can be applied to frozen surfaces, including glass and polypropylene, that have been stored in liquid nitrogen when wrapped around itself by at least 1/8 inches. Matte white polyester material has a permanent acrylic adhesive and is thermal transfer printable. Good print smudge resistance, solvent resistance and high/low temperature performance of -196° to 100°C. This all-in-one cartridge includes label supply and R6400 Series ribbon.
  • Availble in Label Cartridges, or Tape
  • Colour Black on White
  • Brady Material B-492 Ultra Thin Polyester
Specifications
Adhesive TypeAcrylic
ColorBlack/White
MaterialPolyester
Product TypeLabel/Cartridge
For Use With (Equipment)BMP™51, and BMP™53 Label Printers
Quantity240/Pk.
Size12.7 mm dia.
Diameter (Metric)12.7 mm
Unit SizeEach