ColorNatural
Certifications/ComplianceISO 9001:2015; FDA CFR21 GMP; IATA 95 kPa
AutoclavableAutoclavable
Capacity (Metric)50 mL
Bottom ShapeConical
MaterialPolypropylene Resin
Thread StyleExternal
For Use With (Application)Transport Tubes
Length (English)4.53 in.
Length (Metric)115 mm
Packaging10 Tubes / IntegraPack™ bag/5 bags / Pk./2 Pk. / case
Quantity10 Tubes / IntegraPack™ bag
Product LineProtectR
SterilitySterile
DisposableSingle-use
GraduatedYes
Packaging Quantity50
Dnase FreeYes
Closure TypePolyethylene Screw Cap
PrintedYes
Unit SizePack