SuperClear™ 2.0 mL Freestanding Screw Cap Microcentrifuge Tubes
Actual product may vary
Fisherbrand
SuperClear™ 2.0 mL Freestanding Screw Cap Microcentrifuge Tubes
Freestanding tubes with molded graduations and labeling spot.
Have Questions?
Catalog NumberClosure TypePackaging
FKS16674112Attached Cap50 Tubes/Bag, 10 Bags/Pk., 10 Pk./Cs.
Catalog number FKS16674112
Price (SGD)
500.00
Pack
Closure Type:
Attached Cap
Packaging:
50 Tubes/Bag, 10 Bags/Pk., 10 Pk./Cs.
Price (SGD)
500.00
Pack
  • Screw Cap Style Microcentrifuge Tube
  • Freestanding tubes
  • Includes molded graduations and labeling spot
  • ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
  • Polypropylene USP Class VI resin
  • Centrifuging to 20,000 x G
  • Freezable to -80°C; Autoclavable
  • Made in USA
Specifications
Cap StyleScrew Cap
Certifications/ComplianceISO 9001:2015; FDA CFR21 GMP
DescriptionWith attached caps
AutoclavableAutoclavable
Capacity (Metric)2.0 mL
ColorNatural
MaterialPolypropylene Resin
For Use With (Application)Centrifugation
Packaging50 Tubes/Bag, 10 Bags/Pk., 10 Pk./Cs.
Volume (Metric)2.0 mL
Clean ClaimsATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
SterilitySterile
Product LineSuperClear™ microcentrifuge tubes
GraduatedYes
Closure TypeAttached Cap
Max. RCF20,000 RCF
Unit SizePack