Sterileware™ Liquid Thief
Sterileware™ Liquid Thief
Actual product may vary
Sterileware™ Liquid Thief
Sterileware™ Liquid Thief
Sterileware™ Liquid Thief
Fisherbrand
Sterileware™ Liquid Thief
Liquid Thief draws a sample with a simple pull of the handle, then dispenses easily into a vessel.
Have Questions?
Catalog NumberCapacity (Metric)Length (Metric)
FKS16652824100 mL50 cm
Catalog number FKS16652824
Price (SGD)
1,502.00
PK
Add to cart
Capacity (Metric):
100 mL
Length (Metric):
50 cm
Price (SGD)
1,502.00
PK
Add to cart
  • Ideal for sampling water and other low viscosity liquids; 0.15 cm (0.0625 in.) tip hole diameter
  • Made from FDA and EU conforming materials
  • Safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) thief heat sealed in individual polyethylene bag
  • Gamma irradiated sterile (SAL 10-6)
  • Ready for one time use and disposal
  • Lot stamped for accurate tracing
  • Shipped with a Certificate of Processing for Sterilization
Specifications
Product TypeLiquid Sampler
MaterialHDPE
AutoclavableNot Autoclavable
Capacity (English)3.4 oz.
Capacity (Metric)100 mL
SterilitySterile
Quantity20/Pk.
Length (English)19.75 in.
Length (Metric)50 cm
Unit SizePK