Vivaspin™ Turbo 4 Ultrafiltration Unit
Actual product may vary
Fisherbrand
Vivaspin™ Turbo 4 Ultrafiltration Unit
Handles up to 4mL sample volume in 4 swing-out and fixed angle rotors accepting 15mL centrifuge tubes. Sartorius Vivaspin Turbo 4 Ultrafiltration Unit, features twin vertical membranes and sleek internal design for fastest sample concentration with highest recoveries.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMWCOQuantity
FKS1663675210 kDa100/Pk.
FKS1549880110 kDa100/Pk.
FIS14-558-714100 kDa100/Pk.
Catalog number FKS16636752
Price (SGD)
-
MWCO:
10 kDa
Quantity:
100/Pk.
Handles up to 4mL sample volume in 4 swing-out and fixed angle rotors accepting 15mL centrifuge tubes. Sartorius Vivaspin Turbo 4 Ultrafiltration Unit, features twin vertical membranes and sleek internal design for fastest sample concentration with highest recoveries.
  • High flux PES membrane
  • Retentate recovery >95% x pH 1-14 suited polystyrene housing with high chemical compatibility
  • Universal rotor compatibility
  • Volume graduation for easy detection / monitoring of the concentrate volume
  • UV joining technology allows for smooth joint transition between membrane and plastic housing, allowing collection of complete concentrated sample into unique pipette friendly dead stop pocket
Specifications
DescriptionTurbo 4 Ultrafiltration Unit
DisposableSingle-use
Filter TypeUltrafiltration
For Use With (Equipment)Centrifuge
Material (Membrane)PES
MWCO10 kDa
Sample Volume4 mL
SterilityNon-sterile
For Use With (Application)Concentration, Desalting, buffer exchange
Housing MaterialPolystyrene
Max. ForceSwing-out rotor : 4000 x gAngle rotor : 7500 x g
Quantity100/Pk.
Sample TypeSample type: Nanoparticles; Peptide; Protein
TypeCentrifugal Concentrator
Volume (Metric)4 mL
Length (Metric)1225 mm
Unit SizePack