Sterile Antistatic Standard Weighing Boat
Sterile Antistatic Standard Weighing Boat
Actual product may vary
Fisherbrand
Sterile Antistatic Standard Weighing Boat
The Sterile Antistatic Standard Weighing Boat is flexible and features smooth surfaces to provide accurate pour-outs with minimal sample loss.
Have Questions?
Catalog NumberCapacity (Metric)
FKS1620687085 mL
Catalog number FKS16206870
Price (SGD)
312.00
PK
Add to cart
Capacity (Metric):
85 mL
Price (SGD)
312.00
PK
Add to cart
  • Sterile, Antistatic
  • Boat is shallow with a wide, flat bottom to resist tipping
  • Rounded corners simplify transfer
  • Color contrast to ensure sample is completely removed
  • Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases
  • Temperature range -10 to 70°C (14 to 158°F)
  • Can be used with microwave moisture analyzers
  • FDA/food-grade compliant
  • Disposable
  • Manufactured and packaged in a clean room
  • Easy Tear Packaging
Specifications
Capacity (English)2.9 oz.
Certifications/ComplianceFDA
DescriptionSterile Weighing Boat
AutoclavableNot Autoclavable
Bottom ShapeFlat
Capacity (Metric)85 mL
ColorWhite
DisposableSingle-use
MaterialPolystyrene
ShapeSquare
SterilitySterile
TypeWeighing Boat
For Use With (Application)Weighing
For Use With (Equipment)Balance
Quantity200 Pk. (40 x 5)
SizeMedium
Temperature Range (English)14°F to 158°F
Temperature Range (Metric)-10° C to +70° C
Dimensions (L x W)85 x 85 mm
Dimensions (L x W x H)85 x 85 x 24 mm
Length (English)3.3 in.
Length (Metric)85 mm
Width (English)85 mm
Weight (Metric)3.3 in.
Unit SizePK