Clear Straight Sided Glass Jars with White Polypropylene Caps
Actual product may vary
Fisherbrand
Clear Straight Sided Glass Jars with White Polypropylene Caps
Clear, USP Type III soda-lime glass wide mouth design is great for large solid samples and allow for complete removal of contents. Ideal for soil sampling and environmental applications
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure LinerQuantity
FKS16164437480 mLPulp/Vinyl12
FISFB0291192030 mLPulp/Vinyl48
FKS1615451730 mLPulp/Vinyl48
FISFB0291177360 mLPulp/Vinyl24
FISFB02911775120 mLPulp/Vinyl24
FISFB02911776240 mLPulp/Vinyl12
FKS16104447240 mLPulp/Vinyl12
FISFB02911772480 mLPulp/Vinyl12
FKS16184437960 mLPulp/Vinyl12
Catalog number FKS16164437
Price (SGD)
93.00
Pack
Capacity (Metric):
480 mL
Closure Liner:
Pulp/Vinyl
Quantity:
12
Price (SGD)
93.00
Pack
Clear, USP Type III soda-lime glass wide mouth design is great for large solid samples and allow for complete removal of contents. Ideal for soil sampling and environmental applications
Straight sided walls allow for the complete removal of contentsAvailable in a wide variety of sizes, from 1 to 32 oz.
Specifications
Capacity (Metric)480 mL
Capacity (English)16 oz.
ColorClear
Diameter (Metric) Outer Neck86 mm
Diameter (Metric) Inner Neck70 mm
For Use With (Application)Soil Sampling and Environmental Applications
IncludesCap
Height (Metric) With Closure99 mm
MaterialGlass
Quantity12
Closure ColorWhite
Neck Finish89-400
ClarityClear
Certifications/ComplianceUSP Type III Soda Lime Glass Wide Mouth Design is Great for Large Solid Samples and Allow for Complete Removal of Contents
Closure IncludedYes
MouthWide
ShapeRound
Height (Metric)96 mm
Product TypeStraight-sided Jar
Closure LinerPulp/Vinyl
Closure MaterialPolypropylene
Closure Size89-400 mm
Diameter (Metric)91 mm
Closure TypeCap
Unit SizePack