Check Order Status
Clear Standard Wide Mouth Glass Bottles with White Polypropylene Caps
Actual product may vary
Fisherbrand
Clear Standard Wide Mouth Glass Bottles with White Polypropylene Caps
Clear, Type III soda-lime glass ideal for general, small to large volume storage
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Closure Liner
FKS16104407
120 mL
Pulp/Vinyl
FISFB02911770
240 mL
PTFE/Polyethylene Foam
FISFB02911767
480 mL
PTFE/Polyethylene Foam
3 Options
Catalog number
FKS16104407
Price (SGD)
91.00
PK
Add to cart
Capacity (Metric):
120 mL
Closure Liner:
Pulp/Vinyl
Price (SGD)
91.00
PK
Add to cart
Features a wide mouth opening for easy transfer of contents
Applications include pharmaceutical, chemical, environmental, and food packaging and storing
Manufactured from clear flint glass
Specifications
Capacity (Metric)
120 mL
Capacity (English)
4 oz.
Color
Clear
Diameter (Metric) Outer Neck
45 mm
Diameter (Metric) Inner Neck
35 mm
Height (Metric) With Closure
89 mm
Material
Glass (Type 3)
Quantity
24
Closure Color
White
Neck Finish
48-400
Closure Included
Yes
Mouth
Wide
Height (Metric)
86 mm
Product Type
Standard Wide Mouth Bottle
Closure Liner
Pulp/Vinyl
Closure Material
Polypropylene
Closure Size
48-400 mm
Diameter (Metric)
51 mm