Specifically formulated for use with equipment with minimal temperature control. Supplied in 600mL 'ready to use' packs specially designed to eliminate cleaning of laboratory glassware, which is inherently difficult due to the nature of silicone. These range of standards provide both calibration and verification options for rotational viscometer test equipment.
Have Questions?
Change view
Viscosity:
10.12 mPa.s (25°C)
4.988 mPa.s (25°C)
50.49 mPa.s (25°C)
100.7 mPa.s (25°C)
503.9 mPa.s (25°C)
1003 mPa.s (25°C)
5034 mPa.s (25°C)
12423 mPa.s (25°C)
30036 mPa.s (25°C)
60139 mPa.s (25°C)
100075 mPa.s (25°C)
Catalog number FKS16090500
Price (SGD)
280.00
EA
Add to cart
Viscosity:
10.12 mPa.s (25°C)
Request bulk or custom format
Price (SGD)
280.00
EA
Add to cart
Specifications
PackagingTest in pack container
OdorCharacteristic odor
Viscosity10.12 mPa.s (25°C)
Analyte or Component NamesSilicone fluid
Physical FormLiquid
View more
Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
600mL of product supplied in a ready to use 'test in pack' container
1-year shelf life
Ensures full compliance to international test method protocol
Full traceability to National Standards
Tamper evident security packaging
Applicable to all spindles with the exception of the L1 spindle, further provision should be made when using the L1 spindle
Safer working, less mess, less waste, higher throughput