Stainless Steel Spoon/Spatula Combo Utensils
Stainless Steel Spoon/Spatula Combo Utensils
Actual product may vary
Fisher Catalogue
Stainless Steel Spoon/Spatula Combo Utensils
Use one tool for two jobs. Nickel Electro™ Stainless Steel Spoon/Spatula Combo Utensils measure liquids and powders with the spoon end and divide and separate materials with the spatula end.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)
FKS15977961210 mm
FKS11756597120 mm
FKS11864233210 mm
Catalog number FKS15977961
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
210 mm
Dual-use utensil measures liquids and fine powders with the spoon end; divides and separates materials with the spatula end.
  • Manufactured from stainless steel.
Specifications
Quantity10 Pk.
SterilitySterile
AutoclavableAutoclavable
Capacity (Metric)2 mL
Product TypeSpatula
FinishBright Finish
For Use With (Application)Sterile Applications
Width (Metric)20 mm
Dimensions (L x W)210 x 20 mm
Temperature (Metric) Max-196°C to +550°C
MaterialStainless Steel
Length (Metric)210 mm
Unit SizePK