Analytical Balances
Analytical Balances
Actual product may vary
Analytical Balances
Featuring RS232 connectivity for easy communication, a backlit display and a simple interface for uncomplicated operation, these balances are perfectly designed for your workplace.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityCalibration
FKS1597749062 gExternal
FBS15927500220 gInternal
FKS15927500220 gInternal
Catalog number FKS15977490
Price (SGD)
4,511.00
EA
Add to cart
Capacity:
62 g
Calibration:
External
Price (SGD)
4,511.00
EA
Add to cart
  • Large draftshield with sliding top and side doors
  • Standard RS-232 connectivity for easy data transfer
  • Stainless steel platform
  • Two adjustable feet that allow user to level the balance
  • Up-front level indicator - no need to look in back of balance during leveling process
  • Smart design and durable construction - small footprint which saves on benchtop space while providing a large weighing surface
  • Designed for uncomplicated operation with easy-to-use display and interface
  • Calibration lockout security loop in rear of balance
  • Weighing modes include weighing, parts counting and percent weighing
  • Includes AC adapter
  • Certifications: FCC, CE
  • Two year warranty
Specifications
CalibrationExternal
Certifications/ComplianceGLP with optional printer compliance
Connection TypeRS232
Capacity62 g
Display TypeBacklit LCD
Readability0.0001 g
IncludesAC adapter
Linearity0.0002 g
Min. Sample Weight200 mg
ModelFAS64/E
Repeatability0.0001 g
Stabilization Time4 sec.
TypeAnalytical Balance
Warranty2 Years
Weighing ModesWeighing, Parts Counting, Percent Weighing
Dimensions (D x W x H)317 x 201 x 303 mm
Diameter (Metric) Outer Pan90 mm
Electrical Requirements100/240 V ∼200 mA 50/60 Hz 12/18 VA
Amperage0.5 A