Precision Balances
Precision Balances
Actual product may vary
Fisherbrand
Precision Balances
Featuring RS232 connectivity for easy communication, a backlit display and a simple interface for uncomplicated operation, these balances are perfectly designed for your workplace.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityModelCalibrationDimensions (D x W x H)
FKS15957500420 gFPRS423/EExternal317 x 201 x 303 mm
FBS15967500420 gFPRS423Internal317 x 201 x 303 mm
FBS15957500420 gFPRS423/EExternal317 x 201 x 303 mm
FBS159975002200 gFPRS2202/EExternal317 x 201 x 93 mm
FBS159275104200 gFPRS4202Internal317 x 201 x 93 mm
FBS159175104200 gFPRS4202/EExternal317 x 201 x 93 mm
Catalog number FKS15957500
Price (SGD)
3,370.00
EA
Add to cart
Capacity:
420 g
Model:
FPRS423/E
Calibration:
External
Dimensions (D x W x H):
317 x 201 x 303 mm
Price (SGD)
3,370.00
EA
Add to cart
  • Standard RS-232 connectivity for easy data transfer
  • Stainless steel platform
  • Two adjustable feet that allow user to level the balance
  • Up-front level indicator - no need to look in back of balance during leveling process
  • Smart design and durable construction - small footprint which saves on benchtop space while providing a large weighing surface
  • Large, bright backlit display allows for easy viewing of results
  • Four mechanical keys enable easy operation of basic application modes
  • Weighing modes include weighing, parts counting and percent weighing
  • Includes AC adapter
  • Certifications: FCC, CE
  • Two year warranty
Specifications
CalibrationExternal
Certifications/ComplianceGLP with optional printer compliance
Connection TypeRS232
Capacity420 g
Display TypeBacklit LCD
Readability0.001 g
IncludesAC adapter
Linearity0.002 g
Min. Sample Weight0.82 g
ModelFPRS423/E
Repeatability0.001 g
Stabilization Time2 sec.
TypePrecision Balance with Draft Shield
Warranty2 Years
Weighing ModesWeighing, Parts Counting, Percent Weighing
Dimensions (D x W x H)317 x 201 x 303 mm
Diameter (Metric) Outer Pan120 mm
Electrical Requirements100/240 V ∼200 mA 50/60 Hz 12/18 VA
Amperage0.5 A