Contrad 70 Liquid Detergent
Contrad 70 Liquid Detergent
Actual product may vary
Fisherbrand
Contrad 70 Liquid Detergent
Concentrated liquid detergent. Ideal for laboratory washing by hand, for soaking or for use with an ultrasonic bath. For removing soil, grit, organic contaminants, inorganic residue, chemicals, and solvents from glass, ferrous metal, ceramic and plastics.
Have Questions?
Catalog number FKS15937922
Price (SGD)
-
  • Phosphate- and chlorine-free
  • Free-rinsing
  • Specifications
    FormLiquid
    Volume (Metric)5 L
    Unit SizeEA