Conical Tubes with Screw Cap
Conical Tubes with Screw Cap
Actual product may vary
Conical Tubes with Screw Cap
Conical Tubes with Screw Cap
Fisherbrand
Conical Tubes with Screw Cap
Eppendorf™ Conical Tubes with Screw Cap offer reliable sample containment and leak-free storage. SafeCode variants enhance sample identification, while the BioBased options, manufactured using renewable resources, provide an eco-friendly alternative without compromising performance.
Have Questions?
Catalog NumberCapacity (Metric)ColorPurity or Quality GradePackaging
FKS1591518015 mLAmber Sterile, pyrogen-,Dnase-,Rnase-, Human and Bacterial DNA-free200 Tubes (4 Bags x 50 Tubes)
Catalog number FKS15915180
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
15 mL
Color:
Amber
Purity or Quality Grade:
Sterile, pyrogen-,Dnase-,Rnase-, Human and Bacterial DNA-free
Packaging:
200 Tubes (4 Bags x 50 Tubes)
BioBased (made of bio-based material) variant aids in the reduction of consumable-related carbon footprint in the lab. The bio-based tubes have identical material properties and technical specifications as our non-BioBased tubes.
  • Ensures leak-free storage and protects sample integrity.
  • Suited for a wide range of laboratory tasks, enhancing flexibility.
  • High-quality materials for long-lasting, robust performance.
  • Free of plasticizers, biocides or mold release agents.
  • Clear, visible graduations for quick volume assessment.
  • Facilitates optimal sample recovery and efficient centrifugation.
  • Available in 15 mL, 25 mL, and 50 mL formats.
  • 2D Datamatrix (SafeCode) option features 2-level coding for safe sample identification. ECC200 error correction function enables safe code reading with up to 25% damaged codes.
Specifications
Closure MaterialHDPE
ColorAmber
Capacity (Metric)15 mL
MaterialPolypropylene
No. per Pack50 Pk.
Packaging200 Tubes (4 Bags x 50 Tubes)
Quantity200 Pk.
SterilitySterile
AutoclavableNot Autoclavable
Clean ClaimsPyrogen, DNase, RNase, human and bacterial DNA free
Bottom ShapeConical
Closure TypeScrew Cap
Diameter (Metric)17 mm
Max. RCF19,500 x g
Purity or Quality GradeSterile, pyrogen-,Dnase-,Rnase-, Human and Bacterial DNA-free
Unit SizePK