Nalgene™ Narrow-Mouth Bottles Made of Teflon™ PFA with Closure
Nalgene™ Narrow-Mouth Bottles Made of Teflon™ PFA with Closure
Actual product may vary
Fisher Catalogue
Nalgene™ Narrow-Mouth Bottles Made of Teflon™ PFA with Closure
Get excellent resistance to temperature, chemicals and corrosion with Thermo Scientific™ Nalgene™ Narrow-Mouth Bottles Made of Teflon™ PFA with Closure, among the most chemical- and corrosion-resistant containers available.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
FKS1591166160 mL20 mm
FKS15911381125 mL38-430 mm
FIS02-923-35L250 mL38-430 mm
FIS02-923-35N1 L38-430 mm
Catalog number FKS15911661
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
60 mL
Closure Size:
20 mm
Get excellent resistance to temperature, chemicals and corrosion with Thermo Scientific™ Nalgene™ Narrow-Mouth Bottles Made of Teflon™ PFA with Closure, among the most chemical- and corrosion-resistant containers available.
  • Withstand -270°C to +250°C (-454°F to +482°F)
  • Inert to virtually all chemicals except molten alkali metals, fluorine at high temperatures and complex halogenated compounds at high temperatures and pressures
  • Packaged individually
  • Autoclavable
  • Leakproof† PFA closure
Includes:
Leakproof, linerless PFA screw caps
Note:
Completely disengage threads or remove cap before autoclaving
Warranty:
90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity (Metric)60 mL
Capacity (English)2 oz.
ColorTranslucent
DescriptionNarrow Mouth Bottle
Diameter (Metric) Inner Neck14 mm
For Use With (Application)Storage, Transportation ​
MaterialTeflon PFA
No. per Case8/Cs.
No. per Pack1 Ea.
SterilityNon-sterile
Temperature Range (Metric)-100°C to 260°C
Closure ColorNatural
Neck Finish20 mm
Certifications/ComplianceCCD 077, SW-CDC-001501, EN-8-1007-22-0912
Closure IncludedYes
AutoclavableAutoclavable
MouthNarrow
ShapeRound
Height (Metric)84 mm
Product TypeNarrow Mouth Bottle
Width (English)1.55 in.
Height (English)3.30 in.
Closure LinerUnlined
Closure MaterialETFE
Closure Size20 mm
Diameter (Metric)39 mm
Closure TypeScrew Cap
Through the Closure DispensingNo
Unit SizePK