Search Thermo Fisher Scientific
Catalog Number | Size | Length (English) |
---|---|---|
FKS15897101 | Medium | 44.9 in. |
FIS19-181-592 | Small | 44.1 in. |
FKS15887101 | Small | 44.1 in. |
FIS19-181-593 | Medium | 44.9 in. |
FIS19-181-594 | Large | 45.7 in. |
FKS15807111 | Large | 45.7 in. |
FIS19-181-595 | X-Large | 46.5 in. |