Motorised Pipette Controller
Motorised Pipette Controller
Actual product may vary
Fisherbrand
Motorised Pipette Controller
A powerful, lightweight and ergonomically designed pipetting controller for all commonly used glass or plastic pipettes
Have Questions?
Catalog number FKS15850053
Price (SGD)
397.00
EA
Add to cart
Price (SGD)
397.00
EA
Add to cart
  • Powerful yet quiet pump
  • Ergonomic design
  • Soft grip push buttons
  • Double safety valve
  • Autoclavable pipette cone
  • Three modes: High, Low and Gravity
  • Switch located on slide part of controller ensuring ease of operation
  • LED indicator lights up in red when battery is low
  • Can be operated continuously for 8 hours
  • Replaceable membrane Filter to protect device from contaminants being drawn into pipette
  • Can be used while it is charging
Specifications
Battery Type1000mAh NiMH batteries
Compatible TipsCone shape
ColorBlue
Grip TypeSoft Push Button
TypePipette Controller
Volume (Metric)0.1 to 200 mL
Filter TypeHydrophobic PTFE membrane filter
Finger StylePush button
For Use With (Equipment)Glass or plastic serological pipettes
IncludesPipette filler, Charger (GB plug), Desktop stand, Wall mount bracket, Two spare membrane filters: 0.20μm and 0.45μm, Instruction manual, Batteries
Number of SpeedsTwo (High and Low, along with Gravity drain)
Recharge Time2 to 3 hours
Electrical Requirements2 x 1.2V / 1000mAh NiMH batteries
Voltage1.2 V
Unit SizeEA