Polypropylene Black Cryo Boxes
Polypropylene Black Cryo Boxes
Actual product may vary
Fisherbrand
Polypropylene Black Cryo Boxes
Manufactured from high quality polypropylene with a robust hinge and safe snap-on lid.
Have Questions?
Catalog NumberHolds
FKS158485321.2 to 2 mL Cryo Tubes
Catalog number FKS15848532
Price (SGD)
107.00
PK
Add to cart
Holds:
1.2 to 2 mL Cryo Tubes
Price (SGD)
107.00
PK
Add to cart
  • Autoclavable and temperature-resistant from -90° to +121°C
  • 81 places, 9 x 9 grid, numerically coded
  • Stackable for safe transport
  • Drain bores at the bottom for dew liquids
  • Removable grid inserts
  • Can also be used for larger vessels by removing the grid inserts
  • 133 x 133 x 52 mm
Specifications
Closure TypeSnap-On Lid
ColorBlack
MaterialPolypropylene
ShapeSquare
TypeCryo Box
For Use With (Application)Safe Transport
Holds1.2 to 2 mL Cryo Tubes
IncludesRobust Hinge with Snap-On Lid
Max. Temperature (Metric)121°C
Min. Temperature (Metric)-90°C
Quantity5/pk
Tube Array9 x 9 grid
Withstand Temperature (Metric)-90°C to +121°C
Dimensions (L x W x H)133 x 133 x 52 mm
Length (Metric)133 mm
Width (Metric)133 mm
Height (Metric)52 mm
Unit SizePK