Visitorspec Safety Glasses, Coated, Antifog
Visitorspec Safety Glasses, Coated, Antifog
Actual product may vary
Visitorspec Safety Glasses, Coated, Antifog
Visitorspec Safety Glasses, Coated, Antifog
Visitorspec Safety Glasses, Coated, Antifog
Ideal for protecting visitors in industrial or medical settings.
Have Questions?
Catalog number FKS15819257
Price (SGD)
109.00
PK
Add to cart
Price (SGD)
109.00
PK
Add to cart
Ideal for protecting visitors in industrial or medical settings.
  • Economical safety spectacles
  • 100% impact-resistant polycarbonate wraparound unilens design with blue temples
  • Clear lens absorbs >99.9% of harmful UV light
  • Fit over prescription eyeglasses
  • Scratch-resistant, Anti-fog, Anti-static
Specifications
ShapeRectangle
Lens CoatingAnti-fog, Anti-static, Scratch-resistant
Certifications/ComplianceMeets ANSI Z87.1-2003 standards.
Lens TintClear
Product TypeSafety Glasses
Frame ColorClear
Temple StyleAdjustable-length; spatula
MaterialPolycarbonate (Lens)
Lens MaterialPolycarbonate
Unit SizePK