Cryo-Protection™ Face Shield
Cryo-Protection™ Face Shield
Actual product may vary
Fisherbrand
Cryo-Protection™ Face Shield
Molded polycarbonate Cryo-Protection™ Face Shield is shaped to hug your chest and face, and pivots up and out of the way when not in use. The padded head gear has a ratchet band and pin-and-lock system for a 2-way adjustable fit.
Have Questions?
Catalog number FKS15801787
Price (SGD)
481.00
PR
Add to cart
Price (SGD)
481.00
PR
Add to cart
  • Injection molded polycarbonate shield
  • Hugs your chest and the sides of your face
  • Pivots up, away from your face when not in use
  • Ratchet band and pin-and-lock systems provides a 2-way
  • Adjustable fit
  • Lightweight design
  • CE Cat. II
  • Impact resistant after being splashed with liquid cryogens
  • Retains optical clarity
  • Not intended for immersion in liquid nitrogen or other cryogenic liquids
Specifications
Lens TypeSplash Protection
Lens CoatingImpact-resistant
ColorClear
Certifications/Compliance (Europe)CE Category II, Meets ANSI Z87.1
For Use With (Application)Handling Cryogenic Liquids
Frame ColorBlack
Lens MaterialPolycarbonate
Frame TypePadded Head Gear with Ratchet Band
Unit SizePR