Stericup™ Quick Release-GP Sterile Vacuum Filtration Systems
Stericup™ Quick Release-GP Sterile Vacuum Filtration Systems
Actual product may vary
Stericup™ Quick Release-GP Sterile Vacuum Filtration Systems
Stericup™ Quick Release-GP Sterile Vacuum Filtration Systems
Stericup™ Quick Release-GP Sterile Vacuum Filtration Systems
Stericup™ Quick Release-GP Sterile Vacuum Filtration Systems
Stericup™ Quick Release-GP Sterile Vacuum Filtration Systems
For sterile filter applications.
Have Questions?
Catalog NumberDiameter (Metric) Membrane
FKS1578031912.5 cm
Catalog number FKS15780319
Price (SGD)
823.00
PK
Add to cart
Diameter (Metric) Membrane:
12.5 cm
Price (SGD)
823.00
PK
Add to cart
Stericup Quick Release-GP Sterile Vacuum Filtration System
Specifications
ColorClear with blue adapter
DisposableSingle-use
Material (Membrane)PES
Pore Size0.22 μm
SterilitySterile
Filtration Area40 cm2
For Use With (Application)Sterilizing tissue culture media, protein and DNA
Temperature (Metric) Max45°C
TypeBottle Top Filter
Volume (Metric)500 mL
Volume (Metric) Hold Up2.4 mL
Volume (Metric) Receiver500 mL
Volume (Metric) Upper500 mL
Height (Metric)263 mm
Diameter (Metric) Membrane12.5 cm
Unit SizePK