Tube Racks
Tube Racks
Actual product may vary
Tube Racks
Tube Racks
Tube Racks
Fisherbrand
Tube Racks
Optimize workflow with Eppendorf™ Tube Racks that are ideal for sample processing, transport, and short- to mid-term sample storage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPositionsFor Use With (Equipment)
FKS15773178480.5 mL Tubes
FKS15741966361.5 and 2.0 mL Tubes
Catalog number FKS15773178
Price (SGD)
300.00
PK
Add to cart
Positions:
48
For Use With (Equipment):
0.5 mL Tubes
Price (SGD)
300.00
PK
Add to cart
  • Stackable within one rack format and also between all rack variants; saves laboratory space when not in use
  • Non-slip silicone feet and a tilt-proof footprint design ensure safe handling
  • Wavelike design: Tube rows are arranged in different heights so that the reaction vessels are easily accessible and clearly identifiable
  • Made of polypropylene (PP) for high chemical and physical resistance
  • Optimized space-saving dimensions due to perfect tube position-to-footprint arrangement without compromising the tube handling
  • High flexibility: holds tubes of different sizes
  • Numbered tube positions support fast and easy orientation and error-free handling
Specifications
Product TypeTest Tube Stand
HoldsPCR Tubes
MaterialPolypropylene
Positions48
StackableYes
For Use With (Equipment)0.5 mL Tubes
No. per Pack2 Pk.
Unit SizePK