Multi-Purpose Clamps
Actual product may vary
Fisherbrand
Multi-Purpose Clamps
OHAUS™ Multi-Purpose Clamps are designed to securely grip and position a variety of laboratory glassware.
Have Questions?
Catalog NumberSizeJaw TypeLength (Metric)Material
FKS15762399Large3-Prong Dual Adjust273 mmStainless Steel
Catalog number FKS15762399
Price (SGD)
556.00
Each
Size:
Large
Jaw Type:
3-Prong Dual Adjust
Length (Metric):
273 mm
Material:
Stainless Steel
Price (SGD)
556.00
Each
  • Extension arms with a 360° rotation allow placement of apparatus at various distances from lab-frame without compromising experiment integrity
  • Swivel and fixed position clamps hold apparatus near the lab-frame or stand, and have a built-in integral holder for attaching to lab-frames or apparatus
  • Available as single or dual-adjust in nickel-plated zinc or stainless steel
  • Non-slip vinyl sleeves, and fiberglass covers for temperatures above 100°C (included with all 2 and 3-prong clamps)
Specifications
TypeMulti-purpose Clamp
MaterialStainless Steel
IncludesNon-slip vinyl sleeves and fiberglass covers
DescriptionClamp, Multi Purpose, CLM-MULTI3DSL
Diameter (Metric)13 mm
Length (English)10.75 in.
Length (English) Arm5.000 in.
Length (Metric)273 mm
Length (Metric) Arm127 mm
SizeLarge
Size (Metric) Grip0 to 102 mm
Jaw Type3-Prong Dual Adjust
Unit SizeEach