Class B Polypropylene Volumetric Flasks with Screw Caps
Class B Polypropylene Volumetric Flasks with Screw Caps
Actual product may vary
Fisherbrand
Class B Polypropylene Volumetric Flasks with Screw Caps
An excellent alternative to glass flasks, these Class B Polypropylene Volumetric Flasks with Screw Caps are designed to tight tolerances in the laboratory.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeNo. per PackTolerance
FKS1576211650 mLGL1860.12 mL
FKS10392552100 mLGL1860.2 mL
Catalog number FKS15762116
Price (SGD)
241.00
PK
Add to cart
Capacity (Metric):
50 mL
Closure Size:
GL18
No. per Pack:
6
Tolerance:
0.12 mL
Price (SGD)
241.00
PK
Add to cart
  • Highly transparent PP construction
  • Available in a range of capacities and thread sizes
  • Meets Class B tolerances per DIN EN ISO 1042
  • Gravimetrically calibrated 'to contain' at 121°C
  • Flasks can tolerate thermal stress up to 60°C without permanent exceeding of limits
  • To preserve markings, cleaning at no higher than 60°C is recommended
  • Provides excellent resistance to chemicals, aldehydes, alkalis and acids
Specifications
CalibrationCalibrated 'to contain' at 20°C
Closure MaterialPolypropylene
ColorTranslucent
DescriptionIndividually calibrated ring-marking
Capacity (Metric)50 mL
MaterialPolypropylene
Height (Metric)150 mm
No. per Pack6
Product TypeVolumetric Flask
Temperature Range (Metric)60°C max.
Tolerance0.12 mL
AutoclavableNot Autoclavable
DisposableReusable
GraduatedNo
Mouth StyleNarrow
SterilityNon-sterile
ClassB
Bottom ShapePlain
Closure TypeScrew
Surface TreatmentUntreated
Volume (Metric) Working50 mL
Closure SizeGL18
Unit SizePK