Macro, General Purpose Cuvettes AS5C-UV (pair)
Actual product may vary
Fisherbrand
Macro, General Purpose Cuvettes AS5C-UV (pair)
General purpose cuvettes/cells for absorbance, transmission, and other measurements in the UV/VIS wavelength range (260 to 2500 nm).
Have Questions?
Catalog NumberPath Length
FKS1575802210 mm
Catalog number FKS15758022
Price (SGD)
-
Path Length:
10 mm
  • Cuvettes for absorbance/transmission measurements
  • UV/VIS range
  • UV quartz glass construction
  • Macro cells
  • Static sampling
  • Fit all standard 10 mm cell holders
  • Can be used with any beam height
  • Supplied with PTFE lids
Specifications
TypeMacro Cuvette
Capacity (Metric)3.5 mL
MaterialUV Quartz
Closure MaterialPTFE
Width (Metric) Interior9.5 mm
IncludesSquare PTFE Lids
Quantity2/Pk, Matched pair
Path Length10 mm
Wavelength Range260 to 2500 nm
Height (Metric)45 mm
Width (Metric)12.5 mm
Unit SizePack