Search Fisher Scientific
| Catalog Number | Color |
|---|---|
| FKS15748177 | Purple, Gray |
The lab mat is ideal for every type of laboratory and more specialized areas such as cold rooms, clean rooms, sterile suites and areas where high sterility is a requirement. It can be used with items such as centrifuges, vortex mixers, hot plates, stirrers, pipettes and so much more. When using the mat with magnetic stirrers there is a designated area with internal metal inserts to keep stir bars from wandering around the working surface.