Qualitix™ Sterilization Pouch
Qualitix™  Sterilization Pouch
Actual product may vary
Fisherbrand
Qualitix™ Sterilization Pouch
Fits any small to mid-size sterilization device and provides easy and effective wrapping of items. Made of robust medical grade paper, they tightly close with self-sealing strip to maintain sterility. Content remains visible through PET/CPP transparent laminated film.
Have Questions?
Catalog NumberDimensions (L x W)
FKS15730679135 x 260 mm
Catalog number FKS15730679
Price (SGD)
117.00
PK
Add to cart
Dimensions (L x W):
135 x 260 mm
Price (SGD)
117.00
PK
Add to cart
  • Robust durable material
  • Quick content identification
  • Large self-sealing strip, easy to close
  • Chevron top seal for easy opening
  • Suitable for steam and EO gas sterilization
  • Two sterilization indicators
  • Five different sizes
  • Conformity to ISO 17665-06, ISO 11135, CE
Specifications
AutoclavableAutoclavable
Certifications/ComplianceISO 17665-06, ISO 11135, CE
Closure TypeSelf-adhesive
MaterialPaper
Dimensions (L x W)135 x 260 mm
Unit SizePK