Upright ULT Freezer Drawer Rack for Microtiter Plates
Upright ULT Freezer Drawer Rack for Microtiter Plates
Actual product may vary
Fisherbrand
Upright ULT Freezer Drawer Rack for Microtiter Plates
Fisherbrand inventory control rack for 96-Well and 384-Well microtiter plates. Fits all standard types of upright ULT freezers.
Have Questions?
Catalog NumberHolds
FKS15719559Plates with Lid (72)
Catalog number FKS15719559
Price (SGD)
537.00
EA
Add to cart
Holds:
Plates with Lid (72)
Price (SGD)
537.00
EA
Add to cart
  • 4 drawers
  • Holds 72 plates with lid or 96 plates without lid
  • Made of high quality stainless steel
Specifications
Product TypeDrawer Rack
HoldsPlates with Lid (72)
MaterialStainless Steel
For Use With (Application)96/384-Well Microtiter Plates
For Use With (Equipment)Upright ULT Freezer
Dimensions (L x W x H)547 x 247 x 138mm
Unit SizeEA